Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Bio-Inspired “Green” Synthesis of Nanomaterials and their Applications
Dr. Siddharth V. Patwardhan
Department of Chemical and Process Engineering
University of Strathclyde, 75 Montrose Street
Glasgow G1 1XJ, UK
Email: Siddharth.Patwardhan@strath.ac.uk
Introduction
Silica is a very popular inorganic nanomaterial used in a wide range of applications including fillers for rubber, catalyst supports, separation media, carriers in food and agriculture, and abrasive/anticaking agents in cosmetics. It is also widely believed to be an important material for biomedical applications for following reasons:
  • Silica has been approved by the FDA as a material that is “Generally Recognized As Safe” and by the EU for use in cosmetics and food additives.1
  • Silica can be synthesized and modified using a number of wellestablished methods to produce a wide range of distinct varieties of silica (see below for details).1-2
  • Silica has excellent biodegradation/dissolution properties3 and dissolved silica is considered beneficial to bones.4
This article provides an introduction to the various types, chemistry, and traditional syntheses of silica, followed by an overview of the synthesis, properties, and applications of bio-inspired silica, including a perspective on potential applications.  
Silica and Synthesis Methods
Although the term “silica” may suggest a single material type, in reality, the term represents a vast number of materials with distinct structures and properties. This includes silicas that are crystalline (minerals), amorphous, porous (both structured and unstructured pores), colloidal, as well as in a gel or other forms.5 However, each material type is differentiated in a number of ways, including through the position of the silicon and oxygen atoms, the sizes of the primary particle, secondary particles and aggregates, porosities, level of condensation/hydration (i.e., presence of silanol groups: Si–OH), and surface chemistry (e.g., distribution of surface silanols and surface charge). These seemingly subtle differences mainly arise from the method of synthesis, precursor chemistry, catalyst, solvent, finishing method (e.g., drying), and storage and can manifest themselves in significant ways.
A wide range of approaches have been developed for the preparation of silica nanomaterials, including mechanical methods (e.g., ball milling and attrition), physical techniques (e.g., sonochemical, sputtering, and microwave-assisted), chemical routes (vapor phase synthesis, precipitation, micelles, solvothermal, and sol-gel), and combinations thereof.6 In the past, we have witnessed three revolutionary technologies for the synthesis of silica, including the invention of “Stober” synthesis for monodisperse nanoparticles (in 1968),7 as well as two methods for the synthesis of mesoporous silica, including MCM-41 (in 1992),8 and SBA- 15 (in 1998).9 The Stober route enabled the synthesis of monodisperse nanoparticles of controlled sizes, typically between 10–500 nm. Mesoporous silica synthesis, on the other hand, allowed access to porous materials with well-defined and tunable pores that has opened up a number of diverse applications. Collectively, these three papers received more than 22,000 citations, confirming their widespread impact. Mesoporous silicas (e.g., MCM-41 and SBA-15), colloidal Stober silicas, and silica gels prepared via sol-gel methods are most relevant to biomedical applications, including toxicity research. Mesoporous silicas, in particular, have been of wide interest due to their high internal surface areas, large pore volumes, and unique customizable mesoporous structure.1-2 However, the process of mesoporous silica synthesis is typically complex, multistep, and energy intensive, involving harsh conditions including extremes of pH, high temperatures, and the use of highly toxic silica precursors.
Bio-inspired Silica
Biology exerts a remarkable level of control over fabrication of more than 60 distinct nanomaterials such as CaCO3 (Aldrich Prod. No. 481807), Fe3O4 (Aldrich Prod. No. 518158), and silica produced under environmentally friendly conditions.10 Biology has identified a number of “clever” green routes to produce nanomaterials, and an understanding of the secrets of biological nanomaterials formation has the potential to lead to materials with novel applications and/or new technologies for nanomaterials production. The biological production of sophisticated nanomaterials encompasses most of the twelve principles of green chemistry and leads to exciting prospects.11
Biogenic silica is highly sophisticated, hierarchically organized, and produced in a controlled fashion by microorganisms, animals, and plants formed under environmentally friendly and all-aqueous (“green”) conditions in ambient temperatures and pH. Biological and biochemical investigations have made substantial progress in understanding the mechanisms of biosilica formation (Figure 1). This process involves various specialized organic molecules such as proteins and enzymes that regulate the chemistry of silica formation in vivo.11a  For example, amines and amine-functionalized proteins have been identified, isolated, and characterized from silica forming microalga, while a family of specialized silicatein proteins have been discovered from a range of silica forming sponges.11a
In order to further investigate molecular interactions between silica and biomolecules isolated from biosilica forming systems, a number of in vitro studies have been undertaken. These investigations involve the use of advanced analytical tools, the use of model systems as well as modelling and simulations (Figure 1). Such in vitro experiments have produced a great deal of information on biosilicification, in addition to developing greener methods for the synthesis of tailorable silicas that are suitable for novel applications.
Scheme showing how the study of biosilica formation can lead to mechanistic understanding and generate applications
Figure 1. Scheme showing how the study of biosilica formation can lead to mechanistic understanding and generate applications. Image adapted from Patwardhan, Chem. Commun. 2011.11a
Properties of Bio-inspired Synthesis and Bio-inspired Silica
Biologically inspired green synthesis provides significant improvements in comparison with existing materials and methods (Table 1). This includes (1) a one-pot, one-step route, offering substantial reductions in time and energy usage; (2) mild and facile processing using non-hazardous, environmentally benign chemicals, and (3) superior control of product properties to facilitate performance optimization. It is evident from Table 1 that mesoporous silica syntheses requires toxic precursors like tetraethoxysilane, long synthesis (2–6 days), high temperatures (>60 °C), and extremes of pH (needing specialized handling). In contrast, bioinspired silica synthesis is far safer, routinely performed on bench-top at room temperature in water, and can be completed in 5 minutes.
During the last ten years, we have seen an exponential growth in research publications that cover biological and biologically inspired silica.11a For example, researchers have unveiled mechanistic secrets of biosilica formation and how certain biomolecules play a crucial role in nanomaterial deposition through their chemistry and architecture. In vitro experiments on silica formation have enabled the development of bio-inspired green routes to silica by utilizing a wide variety of “additives” (analogues of biomolecules).13 These additives include a range of small to polymeric amines, polypeptides, proteins, and enzymes (selected examples are shown in Figure 2, an extensive list can be found in reference 11a). Furthermore, bio-inspired silica synthesis has been shown to be compatible with a wide variety of silica precursors (Figure 3) with unique features.14 As the understanding of the molecular interactions between additives and nanomaterials are becoming more clear, increased  control over the green synthesis of nanomaterials and their properties  will become possible. For example, we have demonstrated the ability  to tune particle size from <100 nm to 400 nm, surface area between  10–600 m2/g, as well as produce tailored morphologies (Figure 4).15  Various other advantages of the bio-inspired synthesis have been  summarized in Table 2.
Structures of selected additives typically used in bio-inspired silica synthesis
Figure 2. Structures of selected additives typically used in bio-inspired silica synthesis.
Chemistries of silica precursors typically used in bio-inspired silica synthesis
Figure 3. Chemistries of silica precursors typically used in bio-inspired silica synthesis.
Examples of the ability of bio-inspired synthesis to control porosity, growth rates (top), and morphologies (bottom three images)
Figure 4. Examples of the ability of bio-inspired synthesis to control porosity, growth rates (top), and morphologies (bottom three images).
Table 1. Comparison of traditional methods with bio-inspired synthesis.12
Reaction Conditions
Material Reagents Solvents Time (hr) T (°C) pH By-products Finishing
Mesoporous silica TEOS, Surfactant Water, Alcohol 48-144 60–100 ~1 or ~10 Alcohol, Calcined surfactant Calcination at 550 °C (6–24 hr)
Bio-inspired silica Silicate & Additive Water 0.08 20 7 NaCl Drying at 100 °C (2 hr)
Table 2. Summary of properties that can be controlled in bio-inspired silica synthesis. Table adapted from Reference 11a.
Property Nature of Control
Synthesis Very slow to rapid (few seconds) precipitation
Morphology Spherical particles, fibres, sheets, films, gels, hollow particles
Porosity 0–1,000 m2 g−1 (amorphous silica)
Composites With biomolecules, drugs, polymers, quantum dots, catalysts, etc.
Applications
Only the basic principles of bio-inspired synthesis have been observed  and potential applications have only been speculated. Examples of such  potential applications are described below.
Biocatalysis
Bio-inspired routes for catalysis offer significant potential. Key advantages  of this approach include mild processing conditions, facile, rapid and  one-step procedures, and the potential to achieve high enzymatic activity  and stability. This is a “doubly” green approach, since enzymatic processes  are known to provide greener alternatives, while green immobilization  methods eliminate further waste and pollution. A wide range of enzymes  have been successfully immobilized on bio-inspired silica, including  catalase, peroxidases, oxidases, lipases, and many others.16  
The success of biocatalysis was demonstrated using lipase as a commonly  used model of an industrially important enzyme.12 Here, very high levels  of lipase immobilization efficiencies (close to 100%) were reported. It  was revealed that the enzyme retained its full activity; any apparent  reduction in activity was due to mass transfer limitations and not  enzyme deactivation. An important feature of bio-inspired synthesis,  as illustrated in Figure 4, is the ability to tailor physical properties. This  control over physical properties can be exploited to modulate biocatalyst  performance. The enzymes immobilized using bio-inspired synthesis  generally exhibit excellent reuse potential, very low levels of leaching,  improved chemical and thermal stabilities, and sustained performance in  continuous reactor operations. These properties enable their commercial  utilization and, in particular, open up applications in the food and  pharmaceuticals sectors.
Drug Delivery
The mild processing and biocompatibility of silica has led scientists to  explore the ability of bio-inspired silica as a delivery vehicle for carrying  cargo such as fluorescent dyes and drug molecules.11a Although this is  an emerging sector for applications of bio-inspired silica, one example  stands out. Sano et al. reported the loading and successful release of  an anticancer drug using bio-inspired silica.17 It was postulated that  drug release occurs via hydrolysis and disintegration of silica in aqueous  solutions; thus, slowly releasing the anticancer protein resulting in cancer  cell death. The ability to control the chemical properties such as degree of  hydration will prove important in the future exploitation of silica for use in  drug delivery systems.
Environmental Decontamination
Bio-inspired silica is typically micro- and mesoporous, and its porosity can  be tuned by careful control of synthetic conditions. These properties have  been exploited in selective adsorption and removal of pollutants from  air, and by extension, from water as well. For example, the extraction of  formaldehyde from contaminated air streams has been reported using  bio-inspired silicas. The method exploits the selective chemisorption of  formaldehyde by silicas.18 The performance of this approach can easily  rival existing sorbent technologies. When considering the synthesis cost  and time, as well as green implications when preparing silica materials, it is clear that bio-inspired silicas have the potential to provide an exciting  new platform for adsorption and environmental remediation of volatile  organic compounds.
Scalable Manufacturing and Future Applications
Scalability of the synthesis process is crucial to establish the viability of  the industrial-scale production of bio-inspired silica. The scaling-up of a  bench-top reaction is often not trivial for reasons such as the non-linear  nature of both the scaling of laws governing transport phenomena  (e.g., mixing) and the surface area-to-volume ratios. Furthermore,  heterogeneous reactions (i.e., those commonly used for nanomaterial  production) and most unit operations are scale-dependent. Recently, it  has been reported that the synthesis of bio-inspired silica is scalable19  using a bench-top continuous flow system to achieve continuous  production of bio-inspired silica. The ability of this process to manufacture  catalysts and biocatalysts was also reported. Taking it further, a design for  large-scale bio-inspired silica production (~40,000 tonnes per annum) was  recently completed.20
Detailed evaluation of the economic feasibility of this design and a  comparison with existing industrial silica production processes were also  conducted. This feasibility work presented a promising plant design for  large-scale manufacturing of bio-inspired silica. This process would be  significantly economical compared to current production; the payback  period is reduced to 25% of that of the traditional process. Hence, the bioinspired  process shows great promise for industrial-scale development.  Furthermore, the green process was estimated to reduce the  manufacturing carbon footprint by over 90%, mainly by reduced energy  requirements. These benefits, in addition to the potential for improved  control of nanomaterial properties using bio-inspired processes, promises  a very interesting future for the development of bio-inspired silica. 
Materials
References
  1. Garcia-Bennett, A. E. Nanomedicine 2011, 6, 867.
  2. (a) Tang, F.Q.; Li, L.L.; Chen, D. Adv. Mater. 2012, 24, 1504; (b) Brinker, C.J.; Scherer, G.W. Sol-Gel Science: The Physics and Chemistry of Sol-Gel Processing; Academic Press: Boston, 1990.
  3. (a) He, Q.J.; Shi, J.L.; Zhu, M.; Chen, Y.; Chen, F. Micropor. Mesopor. Mater. 2010, 131, 314; (b) Patwardhan, S.V.; Tilburey, G.E.; Perry, C. C. Langmuir 2011, 27, 15135.
  4. Carlisle, E.M. Science 1970, 167, 279.
  5. (a) Iler, R.K. The Chemistry of Silica; John Wiley & Sons: New York, 1979; (b) Kendall, T. Industrial Minerals 2000, March, 49.
  6. (a) Gleiter, H. Prog. Mater. Sci. 1989, 33, 223; (b) Knauth, P.; Schoonman, J. Nanostructured materials; Kluwer Academic Publishers: Boston, MA, 2002.
  7. Stober, W.; Fink, A.; Bohn, E. J. Colloid Interface Sci. 1968, 26, 62.
  8. Kresge, C.T.; Leonowicz, M.E.; Roth, W.J.; Vartuli, J.C.; Beck, J.S. Nature 1992, 359, 710.
  9. Zhao, D.Y.; Feng, J.L.; Huo, Q.S.; Melosh, N.; Fredrickson, G.H.; Chmelka, B.F.; Stucky, G.D. Science 1998, 279, 548.
  10. Biomineralization; Mann, S.; Webb, J.; Williams, R. J. P., Eds.; VCH: Weinheim, 1989.
  11. (a) Patwardhan, S.V. Chem. Commun. 2011, 7567; (b) Anastas, P. T.; Kirchhoff, M. M. Acc. Chem. Res. 2002, 35, 686.
  12. Forsyth, C.; Patwardhan, S.V. J. Mater. Chem. B 2013, 1, 1164.
  13. Patwardhan, S.V.; Clarson, S.J.; Perry, C.C. Chemical Communications 2005, 1113.
  14. Patwardhan, S.V.; Raab, C.; Huesing, N.; Clarson, S.J. Silicon Chem. 2005, 2, 279.
  15. Belton, D.J.; Patwardhan, S.V.; Annenkov, V.V.; Danilovtseva, E.N.; Perry, C.C. Proceedings of the National Academy of Sciences of the United States of America 2008, 105, 5963.
  16. Forsyth, C.; Patwardhan, S.V. In Bio-Inspired Silicon-Based Materials; Zelisko, P.M., Ed.; Springer: in press.
  17. Sano, K.I.; Minamisawa, T.; Shiba, K. Langmuir 2010, 26, 2231.
  18. Ewlad-Ahmed, A.M.; Morris, M.A.; Patwardhan, S.V.; Gibson, L.T. Environ. Sci. Technol. 2012, 46, 13354.
  19. Patwardhan, S.V.; Perry, C.C. Silicon 2010, 2, 33.
  20. Drummond, C.; McCann, R.; Patwardhan, S.V. Chem. Eng. J. 2014, 244, 483.
Related Links