Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Functional Polymers for Biomedical Applications
By: Gangadhar Panambur, Donghun Koo, Philip Dimitrov, Shahinur Rehman, Bryce Nelson, Viktor Balema, SoftMaterials@sial.com
Wide range of functional polymers for biomedical applications have been synthesized and structurally characterized. Several classes of polymers including biodegradable polymers, hydrophilic & amphiphilic polymers, and stimuli responsive polymers have been prepared using controlled and directed functionalization via "living" polymerization such as RAFT, ionic and ring opening polymerization. Selected polymers have been studied for their structure-properties relationship.
Functional Polymers
Polymers represent the leading and versatile type of biomaterials being extensively applied in biomedical applications such as tissue engineering, nano-medicine, medical devices etc.
The versatility of polymers for biomedical uses is attributed to the relative ease with which polymers can be designed with a range of architecture, functionality and appropriate physical, chemical and biological properties.
Functional polymers are special class of polymers where the polymer chains contain one or more reactive groups attached at the chain end or to the backbone. 
In biomedical applications, functional polymers are used as building blocks to design complex structures such as nanoparticles, bioconjugation, biocompatible surfaces, scaffolds, hydrogels etc.
Functionalized Biodegradable Polylactides
Click on image for larger view.
Functionalized Biodegradable Polylactides
Polyoxazolines and Linear polyethylenimines
Polyoxazolines and Linear polyethylenimines
Functionalized Heat & pH Sensitive PNIPAM
Click on image for larger view.
Functionalized Heat & pH Sensitive PNIPAM
Functional RAFT Polymers
Click on image for larger view.
Functional RAFT Polymers
Acknowledgements
Prof. Scott Grayson, Tulane University
Prof. Nicholas L. Abbott, University of Wisconsin