Al(OH)(BDC) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare Al(OH)(BDC). 
Applications
  • Catalysis: Isomerization of α-pinene oxide
Properties
  • Maximum Surface Area: 950 m2 / g BET surface area