Fe-BTC (Basolite F-300) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare Fe-BTC (Basolite F-300). 
Applications
  • Catalysis:  Rearrangement of α-pinene oxide to camphonelal and isopinocamphone
Properties
  • Maximum Surface Area: 1040 m2 / g BET surface area