BIF-11 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare BIF-11. 
Applications
  • Gas adsorption:  Carbon dioxide, nitrogen, hydrogen gas
Properties
  • Unit Cell Volume = 4341.76 Å3