CdIF-1 Supplemental Information
Literature suggests that the following materials can be used to prepare CdIF-1.
- Linker: HMIM, 2-MIM (2-methylimidazole)
- Metal Source: Cadmium Salts (Cd)
- Crytallographic and supplementary information
Properties
- Maximum Surface Area: 2420 m2 / g Langmuir Surface Area
- Pore volume = 0.86 cm3 / g