Co-MOF-74 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare Co-MOF-74. 
Applications
  • Gas adsorption:  Hydrogen, ammonia, cyanogen chloride, sulfur dioxide gas
Properties
  • Maximum Surface Area: 1360 m2 / g BET surface area
  • One Dimensional Pores, Open Metal Sites
Materials
References
  1. Glover, T.; Peterson, G.; Shindler, B.; Britt, D.; Yaghi, O. Chemical Engineering Science 2011, 66(2), 163-170.
  2. Arean, C.; Chavan, S.; Cabello, C.; Garrone, E.; Palomino, G. ChemPhysChem 2010, 11, 3237-3242.
  3. Caskey, S. R., Matzger, A. Material Matters 2009, 4.4, 11.
  4. Tranchemontagne, D. J.; Hunt, J. R.; Yaghi, M. M. Tetrahedron 2008, 64, 8553.
  5. Botas, J.; Calleja, G.; Sanchez-Sanchez, M; Orcajo, G. International Journal of Hydrogen Energy 2011, 36(17), 10834-10844.
Related Links