Co3(BTC)2·12H2O Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare Co3(BTC)2·12H2O. 
Applications
  • Liquid adsorption:  Water, ammonia
Properties
  • Density = 1.946 g / cm3