Cr3(BTC)2 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare Cr3(BTC)2
Applications
  • Separations:  Nitrogen, oxygen gas
Properties
  • Maximum Surface Area: 2040 m2 / g Langmuir Surface Area