Cu-TPTC Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare Cu-TPTC. 
  • Linker: H4TPTC ([1,1':4',1"]Terphenyl-3,3',5,5'-tetracarboxylic acid)
  • Metal Source:   Copper Salts (Cu)
Applications
  • Gas adsorption:  Hydrogen gas
Properties
  • Maximum Surface Area: 2247 m2 / g BET surface area
Materials
References
  1. Lin, X.; Jia, J.; Zhao, X.; Thomas, M.; Blake, A. J.; Walker, G. S.; Champness, N. R.; Hubberstey, P.; Schroder, M.; Angew. Chem. Int. Ed. 2006, 45(44), 7358.
Related Links