Cu(BDC-OH) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare Cu(BDC-OH). 
Applications
  • Gas adsorption:  Carbon dioxide, nitrogen, hydrogen gas
Properties
  • Maximum Surface Area: 397 m2 / g BET surface area
  • Pore volume = 0.214 cm3 / g