DUT-6 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare DUT-6. 
Applications
  • Gas adsorption:  Methane gas, n-butane
  • Liquid Adsorption
  • Separations
Properties
  • Pore volume = 2.02 cm3 / g
Materials
References
  1. Klein, N.; Senkovska, I.; Gedrich, K.; Stoeck, U.; Henschel, A.; Mueller, U.; Kaskel, S. Angew. Chem. Int. Ed. 2009, 48(52), 9954-9957.
Related Links