Gd(TATB)(H2O) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare Gd(TATB)(H2O). 
Applications
  • Gas adsorption:  Hydrogen, nitrogen, and carbon dioxide gas
  • Luminescence