HKUST-1 (Basolite C-300) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare HKUST-1 (Basolite C-300). 
Applications
  • Gas adsorption:  Carbon dioxide, nitrogen, oxygen, hydrogen gas, n-butane
  • Catalysis
  • Separations
Properties
  • Maximum Surface Area: 1958 m2 / g Langmuir Surface Area