Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
IM-22 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare IM-22. 
Applications
  • Separations:  Paraffin isomers
Properties
  • Maximum Surface Area: 575 m2 / g BET surface area
  • Pore volume = 0.24 cm3 / g