IRMOF-8 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare IRMOF-8. 
Applications
  • Catalysis: Friedel-Crafts acetylation
  • Gas adsorption
Properties
  • Maximum Surface Area: 1801 m2 / g BET surface area
  • Pore volume = 0.693 cm3 / g