MOF-74-Mg Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare MOF-74-Mg. 
Applications
  • Gas adsorption:  Carbon dioxide, methane gas
  • Catalysis
Properties
  • Maximum Surface Area: 1525 m2 / g BET surface area
  • One Dimensional Pores, Open Metal Sites
Materials
References
  1. Glover, T.; Peterson, G.; Shindler, B.; Britt, D.; Yaghi, O. Chemical Engineering Science 2011, 66(2), 163-170. 
  2. Bao, Z.; Yu, L.; Ren, Q.; Lu, X.; Deng, S. Journal of Colloid and Interface Science 2011, 353(2), 549-556. 
  3. Rosi, N.; Kim, J.; Eddaoudi, M.; Chen, B.; O'Keeffe, M.; Yaghi, O. J. Am. Chem. Soc. 2005, 127(5), 1504-1518. 
  4. Tranchemontagne, D. J.; Hunt, J. R.; Yaghi, M. M. Tetrahedron 2008, 64, 8553.
Related Links