Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
MgDOBDC Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare MgDOBDC. 
Applications
  • Gas adsorption:  Carbon dioxide gas
Properties
  • Maximum Surface Area: 950 m2 / g BET surface area
Materials
References
  1. Liu, J.; Benin, A.; Furtado, A.; Jakubczak, P.; Willis, R.; LeVan, M. Langmuir 2011, 27(18), 11451-11456.
Related Links