Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
MIL-100 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare MIL-100. 
Applications
When constructed with:
Cr
  • Drug delivery materials:  Ibuprofen
Al
  • None
Properties
When constructed with:
Cr
  • Maximum Surface Area: 3340 m2 / g Langmuir surface area
  • Pore volume = 1.16 cm3 / g
Al
  • None
References
  1. Horcajada, P.; Serre, C.; Vallet-Regi, M.; Sebban, M.; Taulelle, F.; Férey, G. Angew. Chem. Int. Ed. 2006, 45(36), 5974-5978.
  2. Khan, N.; Lee, J.; Jeon, J.; Jhung, S. Microporous and Mesoporous Materials 2012152, 235-239.