MIL-100(Cr) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare MIL-100(Cr). 
Applications
  • Drug delivery materials:  Ibuprofen
Properties
  • Maximum Surface Area: 3340 m2 / g Langmuir surface area
  • Pore volume = 1.16 cm3 / g