MIL-103 Supplemental Information
Literature suggests that the following materials can be used to prepare MIL-103.
- Linker: H3BTB (1,3,5-tris(4-carboxyphenyl)benzene)
- Metal Source: Terbium Salts (Tb)
Applications
- Catalysis
- Luminescence
Properties
- Maximum Surface Area: 930 m2 / g BET surface area
- Pore Diameter = 10 Å