MIL-53(Al) (Basolite A100) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare MIL-53(Al) (Basolite A100). 
Applications
  • Gas adsorption: Hydrogen gas
Properties
  • Maximum Surface Area: 1590 m2 / g Langmuir surface area