MIL-78/MIL-78 (Y, Eu) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare MIL-78. 
Applications
  • Luminescence:  Photoluminescence
Properties
  • Unit Cell Volume = 825 Å3
  • Density: 0.126 g/cm3