MIL-88C-Fe Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare MIL-88C-Fe. 
Applications
  • Catalysis
Properties
  • Maximum Surface Area: 4030 m2 / g BET surface area
  • Unit Cell Volume = 2020 Å3