Mn(BDC)(H2O)2 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare Mn(BDC)(H2O)2
Applications
  • Magnetic resonance imaging
Properties
  • Longitudinal Relaxivity = 5.5
  • Transverse Relaxivity = 80.0 mM-1 s-1