MnSO - MOF Supplemental Information
Literature suggests that the following materials can be used to prepare MnSO - MOF.
- Linker: H4TCPB (1,2,4,5-Tetrakis(4-carboxyphenyl)benzene)
- Metal Source: Zinc Salts (Zn) & Zinc SBU
- Metal Source: Manganese Salts (Mg)
Applications
- Catalysis: size-selective catalysis (epoxidation)
Properties
- Channels = 22.4 Å × 11.7 Å