Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
MOF-1 (Yb-MOF) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare MOF-1 (Yb-MOF). 
Applications
  • Luminescence
  • Sensors
Properties
  • One-dimensional square channels = 7.2 × 7.2 Å
  • Solvent-accessible volume = 1865.2 Å3