MOF-177 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare MOF-177. 
Applications
  • Gas adsorption:  Carbon dioxide, nitrogen, hydrogen gas, large organic molecules
  • Separations
Properties
  • Maximum Surface Area: 5000 m2 / g BET surface area
  • Pore volume = 1.59 cm3 / g