MOF-2 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare MOF-2. 
Applications
  • Separations: Amine exchangability
Properties
  • Maximum Surface Area: 270 m2 / g Langmuir surface area
  • Pore volume = 0.094 cm3 / g