MOF-5 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare MOF-5. 
Applications
  • Gas adsorption: Hydrogen, nitrogen gas
  • Catalysis
Properties
  • Maximum Surface Area: 4400 m2 / g Langmuir surface area