NiDOBDC Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare NiDOBDC. 
Applications
  • Gas adsorption:  Xenon, krypton gas, carbon dioxide gas
Properties
  • Maximum Surface Area: 950 m2 / g BET surface area
Materials
References
  1. Thallapally, P.; Grate, J.; Motkuri, R. Chem. Commun. 2012, 48, 347-349. 
  2. Liu, J.; Benin, A.; Furtado, A.; Jakubczak, P.; Willis, R.; LeVan, M. Langmuir 2011, 27(18), 11451-11456.
Related Links