Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
NOTT-101 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare NOTT-101. 
  • Linker: H4TPTC ([1,1':4',1"]Terphenyl-3,3',5,5'-tetracarboxylic acid)
  • Metal Source:   Copper Salts (Cu)
Applications
  • Gas adsorption:  Hydrogen gas
Properties
  • Maximum Surface Area: 2316 m2 / g BET surface area
Materials
References
  1. Lin, X.; Telepeni, I.; Blake, A. J.; Dailly, A.; Brown, C. M.; Simmons, J. M.; Zoppi, M.; Walker, G. S.; Thomas, M.; Mays, T. J.; Hubberstey, P.; Champness, N. R.; Schroder, M.; J. Am. Chem. Soc. 2009, 131(6), 2159.
Related Links