PCN-17 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare PCN-17. 
Applications
When constructed with:
Dy, Er, Y, Yb
  • Gas adsorption:  Hydrogen gas
Properties
When constructed with:
Dy
  • Maximum Surface Area: 738 m2 / g BET surface area, carbon dioxide gas
  • Pore volume = 0.31 cm3 / g
Er
  • Maximum Surface Area: 606 m2 / g BET surface area, carbon dioxide gas
  • Pore volume = 0.25 cm3 / g
Y
  • Maximum Surface Area: 814 m2 / g BET surface area, carbon dioxide gas
  • Pore volume = 0.33 cm3 / g
Yb
  • Maximum Surface Area: 820 m2 / g BET surface area, carbon dioxide gas
  • Pore volume = 0.34 cm3 / g
Materials
References
  1. Ma, S.; Yuan, D.; Wang, X.; Zhou, H. Inorg. Chem. 2009, 48(5), 2072-2077.
Related Links