PCN-17(Dy) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare PCN-17(Dy). 
Applications
  • Gas adsorption:  Hydrogen gas
Properties
  • Maximum Surface Area: 738 m2 / g BET surface area, carbon dioxide gas
  • Pore volume = 0.31 cm3 / g