PCN-17(Y) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare PCN-17(Y). 
  • Linker: TATB (4,4',4"-s-triazine-2,4,6-triyl-tribenzoic acid)
  • Metal Source: Yttrium Salts (Y)
Applications
  • Gas adsorption:  Hydrogen gas
Properties
  • Maximum Surface Area: 814 m2 / g BET surface area, carbon dioxide gas
  • Pore volume = 0.33 cm3 / g
Materials
References
  1. Ma, S.; Yuan, D.; Wang, X.; Zhou, H. Inorg. Chem. 2009, 48(5), 2072-2077.
Related Links