PCN-19 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare PCN-19. 
Applications
  • Gas adsorption:  Hydrogen gas
Properties
  • Maximum Surface Area: 723 m2 / g BET surface area