PCN-6' Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare PCN-6'. 
  • Linker: TATB (4,4',4"-s-triazine-2,4,6-triyl-tribenzoic acid)
  • Metal Source:   Copper Salts (Cu)
Applications
  • Gas adsorption:  Hydrogen gas
Properties
  • Maximum Surface Area: 3800 m2 / g Langmuir Surface Area
  • Pore volume = 1.453 cm3 / g
Materials
References
  1. Ma, S.; Sun, D.; Ambrogio, M.; Fillinger, J.; Parkin, S.; Zhou, H. J. Am. Chem. Soc. 2007, 129(7), 1858-1859.
Related Links