Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
PCN-9 (Fe) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare PCN-9 (Fe). 
Applications
  • Gas adsorption:  Hydrogen gas
Properties
  • Maximum Surface Area: 682 m2 / g BET surface area
  • Pore volume = 0.33 cm3 / g
Materials
References
  1. Ma, S.; Yuan, D.; Chang, J.; Zhou, H. Inorg. Chem. 2009, 48(12), 5398-5402.
Related Links