Tb-BTC Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare Tb-BTC. 
Applications
  • Luminescence:  Photoluminescence
Properties
  • Maximum Surface Area: 678 m2 / g BET surface area
  • Pore volume = 0.35 cm3 / g