Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Tb(TATB)(H2O) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare Tb(TATB)(H2O). 
  • LinkerTATB (4,4',4"-s-triazine-2,4,6-triyl-tribenzoic acid)
  • Metal Source:   Terbium Salts (Tb)
Applications
  • Gas adsorption:  Hydrogen, nitrogen, and carbon dioxide gas
  • Luminescence
Materials
References
  1. Zhang, H.; Li, N.; Tian, C.; Liu, T.; Du, F.; Lin, P.; Li, Z.; Du, S. Crystal Growth & Design 2012, 12(2), 670-678.
Related Links