TIF-A1 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare TIF-A1. 
Applications
  • Gas adsorption:  Carbon dioxide gas
Properties
  • Maximum Surface Area: 718 m2 / g Langmuir Surface Area