TO - MOF Supplemental Information
Literature suggests that the following materials can be used to prepare TO - MOF.
- Linker: H4TCPB (1,2,4,5-Tetrakis(4-carboxyphenyl)benzene)
- Metal Source: Zinc Salts (Zn) & Zinc SBU
Applications
- Gas adsorption
Properties
- Maximum Surface Area: 512 m2 / g BET surface area, carbon dioxide gas