TUDMOF-1 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare TUDMOF-1. 
Applications
  • Gas adsorption:  Hydrogen gas
  • Magnetism
Properties
  • Maximum Surface Area: 1280 m2 / g BET surface area
  • Pore volume = 0.67 cm3 / g