UiO-66(Zr) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare UiO-66(Zr). 
Applications
  • Gas adsorption:  Carbon dioxide, methane gas
  • Separations
Properties
  • Maximum Surface Area: 1067 m2 / g BET surface area
  • Pore volume = 0.40 cm3 / g