UMCM-1 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare UMCM-1. 
Applications
  • Gas adsorption:  Nitrogen gas
  • Catalysis
Properties
  • Maximum Surface Area: 4160 m2 / g BET surface area
Materials
References
  1. Koh, K.; Wong-Foy, A.; Matzger, A. Angew. Chem. Int. Ed. 2008, 47(4), 677-680.
Related Links