Y(TATB)(H2O) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare Y(TATB)(H2O). 
  • LinkerTATB (4,4',4"-s-triazine-2,4,6-triyl-tribenzoic acid)
  • Metal Source:   Yttrium Salts (Y)
Applications
  • Gas adsorption:  Hydrogen, nitrogen, and carbon dioxide gas
  • Luminescence
Materials
References
  1. Zhang, H.; Li, N.; Tian, C.; Liu, T.; Du, F.; Lin, P.; Li, Z.; Du, S. Crystal Growth & Design 2012, 12(2), 670-678.
Related Links