ZIF-8 (Basolite Z1200) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare ZIF-8 (Basolite Z1200). 
Applications
When constructed with:
Zn
  • Separations:  Paraffin isomers, alkanes
  • Gas Adsorption
Zn (Basolite Z1200)
  • Gas Adsorption:  Hydrogen gas
  • Sensors
Properties
When constructed with:
Zn
  • Maximum Surface Area: 1813 m2 / g BET surface area
  • Pore volume = 0.65 cm3 / g
Zn (Basolite Z1200)
  • Maximum Surface Area: 1810 m2 / g Langmuir Surface Area
  • Pore volume = 0.636 cm3 / g