ZnPO - MOF Supplemental Information
Literature suggests that the following materials can be used to prepare ZnPO - MOF.
- Linker: H4TCPB (1,2,4,5-Tetrakis(4-carboxyphenyl)benzene)
- Metal Source: Zinc Salts (Zn) & Zinc SBU
Applications
- Catalysis
Properties
- Maximum Surface Area: 3000 m2 / g BET surface area
- Channels (a-axis) = 15 Å × 9 Å
- Channels (b-axis) = 11 Å × 9 Å
- Channels (c-axis) = 8 Å × 9 Å