Surface & Personnel Monitoring: HYCON® Contact Slides
HYCON® Contact Slides - Ready-to-use media for microbial testing of flat and curved surfaces
For easy and flexible microbiological monitoring of surfaces in the food & beverage industry. The unique design of these HYCON® contact slides allows testing on flat, curved or irregular surfaces and makes them a universal tool and ideally suited to the production environment found in food and beverage production.
  • Offered as a range of total count media as well as selective media for yeasts and molds, and for coliform bacteria.
  • Manufactured under controlled aseptic conditions
  • Single packed strips to minimize waste
  • Shelf life of up to six months to help save costs
  • Room temperature storage of nonselective media giving cost savings on temperature controlled storage
  • Rectangular shape (25 cm2)
Materials