Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Heavy Wall CD Vials Increase Sample Recovery
Reporter US Volume 28.3
Heavy wall center drain (CD™) vials are an excellent choice of vials for analysts seeking to increase sample recovery when performing high temperature and/or high-pressure applications.
The CD vial has a unique conical interior bottom that allows the sample to completely drain to the center bottom of the vial. This patented design allows maximum extraction of the sample using a syringe or micropipette, with less than 4 μL of sample being left behind.
These vials are manufactured from 2.5 mm wall borosilicate glass tubing that provides a more durable vessel than standard wall vials for high pressure and/or high temperature applications that include lyophilization, derivatization and other small-scale reactions. This durability allows the heavy wall CD vial to be used as a replacement for V-vials and Micro Reaction vessels that are traditionally used in high temperature applications.
Heavy wall CD vials are also priced 30% less than the comparable V-vials and Micro Reaction vessels providing analysts with a quality alternative at a considerable savings.
Heavy wall CD vials have the following benefits
  • Greater sample recovery
  • Economically priced
  • Autoclavable
  • Temperature range of -70 °C to 160 °C
  • Manufactured from Type 1 borosilicate glass
  • Vial, cap and septa are pre-assembled
The heavy wall CD vials are available in a pack of 12 with a black cap and PTFE/silicone septa.